Learn More
2,6-Dichloro-3-cyano-4-methylpyridine, 97%, Thermo Scientific™
Catalog Number 11404237
Click to view available options
Quantità:
1 g
5 g
25 g
875-35-4
C7H4Cl2N2
187.023
MFCD00052633
LSPMHHJCDSFAAY-UHFFFAOYSA-N
2,6-dichloro-4-methylnicotinonitrile, 2,6-dichloro-3-cyano-4-methylpyridine, 3-cyano-2,6-dichloro-4-methylpyridine, 3-cyano-2,6-dichloro-4-methyl pyridine, 3-pyridinecarbonitrile, 2,6-dichloro-4-methyl, 3-cyano-4-methyl-2,6-dichloropyridine, 2,6-dichloro-4-methyl-3-cyanopyridine, 2,6-dichloro-4-methylnicotinonitrile 3-cyano-2,6-dichloro-4-methylpyridine, pubchem15375, acmc-209qo9
70131
2,6-dichloro-4-methylpyridine-3-carbonitrile
CC1=CC(=NC(=C1C#N)Cl)Cl
875-35-4
C7H4Cl2N2
187.023
MFCD00052633
LSPMHHJCDSFAAY-UHFFFAOYSA-N
2,6-dichloro-4-methylnicotinonitrile, 2,6-dichloro-3-cyano-4-methylpyridine, 3-cyano-2,6-dichloro-4-methylpyridine, 3-cyano-2,6-dichloro-4-methyl pyridine, 3-pyridinecarbonitrile, 2,6-dichloro-4-methyl, 3-cyano-4-methyl-2,6-dichloropyridine, 2,6-dichloro-4-methyl-3-cyanopyridine, 2,6-dichloro-4-methylnicotinonitrile 3-cyano-2,6-dichloro-4-methylpyridine, pubchem15375, acmc-209qo9
70131
2,6-dichloro-4-methylpyridine-3-carbonitrile
CC1=CC(=NC(=C1C#N)Cl)Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2,6-Dichloro-3-cyano-4-methylpyridine
Punto di fusione 108°C to 112°C
Quantità 1 g
Numero UN UN3439
Peso formulazione 187.03
Percent Purity 97%