missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitrotoluene, 99%, Thermo Scientific Chemicals

Catalog Number p-7025164
Click to view available options
Quantità:
5 g
10 g
50 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
29682-46-0
C7H5Cl2NO2
206.02
MFCD00024185
WBNZUUIFTPNYRN-UHFFFAOYSA-N
2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
97155
1,3-dichloro-2-methyl-4-nitrobenzene
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
29682-46-0
C7H5Cl2NO2
206.02
MFCD00024185
WBNZUUIFTPNYRN-UHFFFAOYSA-N
2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
97155
1,3-dichloro-2-methyl-4-nitrobenzene
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O

2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2,6-Dichloro-3-nitrotoluene
Punto di fusione 53°C to 55°C
Quantità 5 g
Numero UN UN2811
Beilstein 2259855
Informazioni di solubilità Insoluble in water.
Peso formulazione 206.03
Percent Purity 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'