Learn More
2,6-Dimethylphenol-d9 (Major), TRC
Product Code. 30384411
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384411 25 mg
30383435 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30384411 Supplier Toronto Research Chemicals Supplier No. TRCD46585725MG
 Restricted Item
Spedizione stimata: 11-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1021325-40-5
Molecular Formula C8HD9O
Molecular Weight (g/mol) 131.22
Synonym 2,6-Xylenol-d9, 1-Hydroxy-2,6-dimethylbenzene-d9, 2,6-Dimethylphenol-d9, NSC 2123-d9
SMILES [2H]C1=C([2H])C(C([2H])([2H])[2H])=C(O)C(C([2H])([2H])[2H])=C1[2H]
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3/i1D3,2D3,3D,4D,5D
Formula Weight 131.1297
Chemical Name or Material 2,6-Dimethylphenol-d9