missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylphenol, 98%, Thermo Scientific Chemicals

Catalog Number p-7047004
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
2432-11-3
MFCD00009716
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
2432-11-3
MFCD00009716

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2,6-Diphenylphenol
Punto di fusione 101°C to 103°C
Formula molecolare C18H14O
Quantità 5 g
Beilstein 2051224
Sinonimo 1,1':3',1-terphenyl-2'-ol, m-terphenyl-2'-ol, 2,6-diphenyl phenol, 2'-hydroxy-m-terphenyl, unii-la93w35m96, 3-phenyl-1,1'-biphenyl-2-ol, 2,6-di-phenylphenol, 2,6,-diphenylphenol, 2,6-di phenyl phenol
InChI Key ATGFTMUSEPZNJD-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=C(C(=CC=C2)C3=CC=CC=C3)O
IUPAC Name 2,6-diphenylphenol
Molecular Weight (g/mol) 246.31
PubChem CID 75512
Peso formulazione 246.31
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'