Learn More
2-Acetamido-4,6-O-benzylidene-2-deoxy-b-D-glucopyranosyl Azide, TRC
Product Code. 30322957
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322957 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30322957 Supplier Toronto Research Chemicals Supplier No. TRCA168103100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl Azide
CAS 168397-51-1
Molecular Formula C15H18N4O5
Quantity 100 mg
InChI Formula InChI=1S/C15H18N4O5/c1-8(20)17-11-12(21)13-10(23-14(11)18-19-16)7-22-15(24-13)9-5-3-2-4-6-9/h2-6,10-15,21H,7H2,1H3,(H,17,20)
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@@H]2O[C@@H](OC[C@H]2O[C@H]1N=[N+]=[N-])c3ccccc3
IUPAC Name N-(6-azido-8-hydroxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl)acetamide
Molecular Weight (g/mol) 334.33
Formula Weight 334.1277