missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone, 98%, Thermo Scientific Chemicals

Catalog Number p-3777186
Click to view available options
Quantità:
5 g
25 g
100 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
700-58-3
C10H14O
150.22
MFCD00074737
IYKFYARMMIESOX-UHFFFAOYSA-N
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
64151
CHEBI:40611
adamantan-2-one
C1C2CC3CC1CC(C2)C3=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
700-58-3
C10H14O
150.22
MFCD00074737
IYKFYARMMIESOX-UHFFFAOYSA-N
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
64151
CHEBI:40611
adamantan-2-one
C1C2CC3CC1CC(C2)C3=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Adamantanone
CAS min. % 97.5
CAS max % 100.0
Punto di fusione 282°C to 286°C
Colore White
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97.5% min. (GC)
Imballaggio Glass bottle
Quantità 5 g
Informazioni di solubilità Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol and chloroform
Peso formulazione 150.22
Percent Purity 98%
Forma fisica Crystalline Powder
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'