Learn More
2-Amino-3-benzyloxypyridine, TRC
Product Code. 30325547
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325547 25 g 25g
30315677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30325547 Supplier Toronto Research Chemicals Supplier No. TRCA60022525G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-3-benzyloxypyridine
CAS 24016-03-3
Molecular Formula C12 H12 N2 O
Quantity 25 g
Synonym 2-Pyridinamine, 3-(phenylmethoxy)-, Pyridine, 2-amino-3-(benzyloxy)- (8CI), 2-Amino-3-(benzoxy)pyridine, 2-Amino-3-benzyloxypyridine, 3-(Phenylmethoxy)-2-pyridinamine, 3-Benzyloxy-2-aminopyridine, 3-Benzyloxypyridin-2-amine, NSC 298538, [(3-(Benzyloxy)pyridin-2-yl])amine
InChI Formula InChI=1S/C12H12N2O/c13-12-11(7-4-8-14-12)15-9-10-5-2-1-3-6-10/h1-8H,9H2,(H2,13,14)
SMILES Nc1ncccc1OCc2ccccc2
IUPAC Name 3-phenylmethoxypyridin-2-amine
Molecular Weight (g/mol) 200.24
Formula Weight 200.095