Learn More
2-Amino-4-hydroxy-6-methylpyrimidine, 99%, Thermo Scientific Chemicals
Catalog Number p-56723
18.06 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.
Click to view available options
Quantità:
25 g
100 g
Imballaggio:
Plastic bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3977-29-5
C5H7N3O
125.13
MFCD00006095
KWXIPEYKZKIAKR-UHFFFAOYSA-N
2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
1532
CHEBI:58959
2-amino-6-methyl-1H-pyrimidin-4-one
CC1=CC(=O)N=C(N1)N
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3977-29-5
C5H7N3O
125.13
MFCD00006095
KWXIPEYKZKIAKR-UHFFFAOYSA-N
2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
1532
CHEBI:58959
2-amino-6-methyl-1H-pyrimidin-4-one
CC1=CC(=O)N=C(N1)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Amino-4-hydroxy-6-methylpyrimidine
Nota nome 99%
CAS min. % 98.5
CAS max % 100.0
Punto di fusione >300.0°C
Colore White to Cream
Spettro a infrarossi Authentic
Intervallo percentuale saggio 99%
Imballaggio Plastic bottle
Quantità 25 g
Beilstein 24, 343
Informazioni di solubilità Solubility in water: insoluble.
Peso formulazione 125.13
Percent Purity 99%
Forma fisica Powder