missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-iodopyridine, 95%, Thermo Scientific™

Catalog Number p-7029576
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
211308-81-5
C5H4ClIN2
254.455
MFCD07644578
DIONPYCYVWCDIG-UHFFFAOYSA-N
2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
11076010
5-chloro-3-iodopyridin-2-amine
C1=C(C=NC(=C1I)N)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
211308-81-5
C5H4ClIN2
254.455
MFCD07644578
DIONPYCYVWCDIG-UHFFFAOYSA-N
2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
11076010
5-chloro-3-iodopyridin-2-amine
C1=C(C=NC(=C1I)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Amino-5-chloro-3-iodopyridine
Punto di fusione 95°C to 98°C
Quantità 250 mg
Sensibilità Light sensitive
Peso formulazione 254.46
Percent Purity 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'