Learn More
2-Amino-N-methylbenzamide, TRC
Specifications
Specifications
| Materiale o nome chimico | 2-Amino-N-methylbenzamide |
| CAS | 4141-08-6 |
| Formula molecolare | C8H10N2O |
| Quantità | 10 g |
| Sinonimo | o-Amino-N-methylbenzamide, Anthranilic Acid Methylamide, N-Methyl-2-aminobenzamide, N-Methylanthranilamide, NSC 207861, [2-[(Methylamino)carbonyl]phenyl]amine, o-Amino-N-methylbenzamide |
| InChI Formula | InChI=1S/C8H10N2O/c1-10-8(11)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3,(H,10,11) |
| SMILES | CNC(=O)c1ccccc1N |
| IUPAC Name | 2-amino-N-methylbenzamide |
| Molecular Weight (g/mol) | 150.18 |
| Peso formulazione | 150.0793 |