missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,6-dimethyl Ester Hexanedioic Acid, TRC

Codice prodotto. 30336165
Cambia vista
Click to view available options
Quantità:
100 mg
Dimensione della confezione:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30336165 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336165 Supplier Toronto Research Chemicals Supplier No. TRCB680695100MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 2-Bromo-1,6-dimethyl Ester Hexanedioic Acid
CAS 3196-19-8
Formula molecolare C16 H18 N O10 S . Na
Quantità 100 mg
InChI Formula InChI=1S/C16H19NO10S.Na/c1-7-4-12(19)26-10-5-8(2-3-9(7)10)25-16-13(17-28(22,23)24)15(21)14(20)11(6-18)27-16;/h2-5,11,13-18,20-21H,6H2,1H3,(H,22,23,24);/q;+1/p-1/t11?,13?,14-,15?,16+;/m0./s1
SMILES [Na+].CC1=CC(=O)Oc2cc(O[C@@H]3OC(CO)[C@H](O)C(O)C3NS(=O)(=O)[O-])ccc12
IUPAC Name sodium;N-[(2S,5R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]sulfamate
Molecular Weight (g/mol) 439.37
Peso formulazione 439.055
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.