Learn More
2-Bromo-2-propylpentanoic Acid Ethyl Ester, TRC
Codice prodotto. 30333162
Cambia vista
Click to view available options
Quantità:
250 mg
Dimensione della confezione:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30333162 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30333162 Fornitore Toronto Research Chemicals N. del fornitore TRCB686720250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 2-Bromo-2-propylpentanoic Acid Ethyl Ester
CAS 99174-91-1
Formula molecolare C10 H19 Br O2
Quantità 250 mg
InChI Formula InChI=1S/C10H19BrO2/c1-4-7-10(11,8-5-2)9(12)13-6-3/h4-8H2,1-3H3
SMILES CCCC(Br)(CCC)C(=O)OCC
IUPAC Name ethyl 2-bromo-2-propylpentanoate
Molecular Weight (g/mol) 251.16
Peso formulazione 250.0568