Learn More
2-Bromo-2'-(trifluoromethyl)acetophenone, 97%
Product Code. 15444707
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444707 5 g 5g
15454707 250 mg 250mg
15434707 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15444707 Supplier Thermo Scientific Alfa Aesar Supplier No. H32032.06
 Restricted Item
Spedizione stimata: 02-12-2026
Add to basket
CAS: 54109-16-9 | C9H6BrF3O | 267.045 g/mol
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54109-16-9
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.045
MDL Number MFCD03094304
InChI Key KWZCBMKXNYOQAK-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenacyl bromide, 2-bromo-1-2-trifluoromethyl phenyl-1-ethanone, 2-bromo-2'-trifluoromethyl acetophenone, 2-bromo-1-2-trifluoromethyl-phenyl-ethanone, 2-bromo-1-2-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2-trifluoromethyl phenyl ethanone, ethanone, 2-bromo-1-2-trifluoromethyl phenyl, 2-trifluoromethyphenacyl bromide, 2-trifluoromethyl phenacylbromide
PubChem CID 2778430
IUPAC Name 2-bromo-1-[2-(trifluoromethyl)phenyl]ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)C(F)(F)F
Specifications
Melting Point 24°C to 32°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 267.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-2'-(trifluoromethyl)acetophenone