Learn More
2-(Bromomethyl)-3-nitrobenzoic Acid Methyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | 2-(Bromomethyl)-3-nitrobenzoic Acid Methyl Ester |
| CAS | 98475-07-1 |
| Formula molecolare | C9 H8 Br N O4 |
| Quantità | 10 g |
| Sinonimo | Benzoic acid, 2-(bromomethyl)-3-nitro-, methyl ester (9CI, ACI), 2-(Bromomethyl)-3-nitrobenzoic acid methyl ester, Methyl 2-(bromomethyl)-3-nitrobenzoate |
| InChI Formula | InChI=1S/C9H8BrNO4/c1-15-9(12)6-3-2-4-8(11(13)14)7(6)5-10/h2-4H,5H2,1H3 |
| SMILES | COC(=O)c1cccc(c1CBr)[N+](=O)[O-] |
| IUPAC Name | methyl 2-(bromomethyl)-3-nitrobenzoate |
| Molecular Weight (g/mol) | 274.07 |
| Peso formulazione | 272.9637 |