Learn More
2-Chloro-1-(4-chlorophenyl)ethanone, TRC
Product Code. 30355168
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355168 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30355168 Supplier Toronto Research Chemicals Supplier No. TRCC368618100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-1-(4-chlorophenyl)ethanone
CAS 937-20-2
Molecular Formula C8H6OCl2
Quantity 100 mg
InChI Formula InChI=1S/C8H6Cl2O/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4H,5H2
SMILES ClCC(=O)c1ccc(Cl)cc1
IUPAC Name 2-chloro-1-(4-chlorophenyl)ethanone
Molecular Weight (g/mol) 189.03
Formula Weight 187.9796