Learn More
2’-Chloro-2-bromoacetophenone, TRC
Product Code. 30354489
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30354489 Supplier Toronto Research Chemicals Supplier No. TRCC3650341G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2'-Chloro-2-bromoacetophenone
CAS 5000-66-8
Molecular Formula C8 H6 Br Cl O
Quantity 1 g
Synonym Tulobuterol Impurity 1
InChI Formula InChI=1S/C8H6BrClO/c9-5-8(11)6-3-1-2-4-7(6)10/h1-4H,5H2
SMILES Clc1ccccc1C(=O)CBr
IUPAC Name 2-bromo-1-(2-chlorophenyl)ethanone
Molecular Weight (g/mol) 233.49
Formula Weight 231.9291