missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-fluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-2698066
Click to view available options
Quantità:
1 g
10 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
85958-57-2
C6H3Cl2FO2S
229.046
MFCD00052957
FCFPJKHVCHKCMP-UHFFFAOYSA-N
2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
522846
2-chloro-4-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
85958-57-2
C6H3Cl2FO2S
229.046
MFCD00052957
FCFPJKHVCHKCMP-UHFFFAOYSA-N
2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
522846
2-chloro-4-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl
Materiale o nome chimico 2-Chloro-4-fluorobenzene-1-sulfonyl chloride
CAS min. % 97.0
Intervallo percentuale saggio 97%
Imballaggio Amber Glass Bottle
Formula lineare Cl(F)C6H3(SO2Cl)
Quantità 1 g
Peso formulazione 229.06
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'