Learn More
2-Chloro-N,N-dimethylethylamine-d6 Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | 2-Chloro-N,N-dimethylethylamine dimethyl-d6 hydrochloride |
| CAS | 97941-91-8 |
| Formula molecolare | C4 2H6 H4 Cl N . Cl H |
| Quantità | 10 mg |
| Sinonimo | Ethanamine, 2-chloro-N,N-di(methyl-d3)-, hydrochloride (1:1) (ACI), Ethanamine, 2-chloro-N,N-di(methyl-d3)-, hydrochloride (9CI), 2-Chloro-N,N-dimethyl-d6-ethylamine HCl, 2-Chloro-N,N-di(methyl-d3)ethanamine hydrochloride, N-(2-Chloroethyl)-N,N-dimethylamine-D6 (N,N-dimethyl-D6) hydrochloride |
| InChI Formula | InChI=1S/C4H10ClN.ClH/c1-6(2)4-3-5;/h3-4H2,1-2H3;1H/i1D3,2D3; |
| SMILES | Cl.[2H]C([2H])([2H])N(CCCl)C([2H])([2H])[2H] |
| IUPAC Name | 2-chloro-N,N-bis(trideuteriomethyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 150.08 |
| Peso formulazione | 149.0645 |