missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetoacetanilide, 98%, Thermo Scientific™

Catalog Number p-7024290
Click to view available options
Quantità:
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
93-70-9
C10H10ClNO2
211.65
MFCD00018224
BFVHBHKMLIBQNN-UHFFFAOYSA-N
2'-chloroacetoacetanilide, n-2-chlorophenyl-3-oxobutanamide, aaoc, o-chloroacetoacetanilide, acetoacet-o-chloroanilide, butanamide, n-2-chlorophenyl-3-oxo, acetoacet-o-chloranilide, o-acetoacetochloroanilide, n-acetoacetyl-2-chloroaniline, acetoacetyl-o-chloroanilide
7156
N-(2-chlorophenyl)-3-oxobutanamide
CC(=O)CC(=O)NC1=CC=CC=C1Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
93-70-9
C10H10ClNO2
211.65
MFCD00018224
BFVHBHKMLIBQNN-UHFFFAOYSA-N
2'-chloroacetoacetanilide, n-2-chlorophenyl-3-oxobutanamide, aaoc, o-chloroacetoacetanilide, acetoacet-o-chloroanilide, butanamide, n-2-chlorophenyl-3-oxo, acetoacet-o-chloranilide, o-acetoacetochloroanilide, n-acetoacetyl-2-chloroaniline, acetoacetyl-o-chloroanilide
7156
N-(2-chlorophenyl)-3-oxobutanamide
CC(=O)CC(=O)NC1=CC=CC=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2'-Chloroacetoacetanilide
Punto di fusione 104°C to 106°C
Quantità 25 g
Peso formulazione 211.65
Percent Purity 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'