missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-3-iodopyridine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantità:
250 mg
1 g
Descrizione
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifica
Materiale o nome chimico | 2-Cyano-3-iodopyridine |
Punto di fusione | 98°C to 100°C |
Quantità | 250 mg |
Numero UN | UN3439 |
Sensibilità | Light sensitive |
Peso formulazione | 230.01 |
Percent Purity | 95% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'