missing translation for 'onlineSavingsMsg'
Learn More

2-Diphenylphosphino-1-naphthoic acid, 97%, Thermo Scientific Chemicals

Codice prodotto. 11436096
Cambia vista
Click to view available options
Quantità:
1 g
5 g
25 g
Dimensione della confezione:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11436096 25 g 25g
11416096 1 g 1g
11426096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11436096 Fornitore Thermo Scientific Alfa Aesar N. del fornitore B22361.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178176-80-2
Formula molecolare C23H17O2P
Molecular Weight (g/mol) 356.36
Numero MDL MFCD02683090
InChI Key WJCBDBBZIGUHOB-UHFFFAOYSA-N
Sinonimo 2-diphenylphosphino-1-naphthoic acid, 2-diphenylphosphanyl naphthalene-1-carboxylic acid, acmc-20anx1, 2-diphenylphosphino-1-naphthalenecarboxylic acid, 2-diphenylphosphino-naphthalen-1-carboxylic acid, 1-naphthalenecarboxylicacid, 2-diphenylphosphino
PubChem CID 10861198
IUPAC Name 2-diphenylphosphanylnaphthalene-1-carboxylic acid
SMILES OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Materiale o nome chimico 2-Diphenylphosphino-1-naphthoic acid
Punto di fusione 190°C to 192°C
Quantità 25 g
Peso formulazione 356.36
Percent Purity 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.