Learn More
2-Ethoxy-6,9-diaminoacridine Lactate, TRC
Product Code. 30372662
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372662 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30372662 Supplier Toronto Research Chemicals Supplier No. TRCE2615055G
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethacridine Lactate
CAS 1837-57-6
Molecular Formula C15 H15 N3 O . C3 H6 O3
Quantity 5 g
Synonym Propanoic acid, 2-hydroxy-, compd. with 7-ethoxy-3,9-acridinediamine (1:1), 6,9-Diamino-2-ethoxyacridine lactate, Lactic acid compd. with 6,9-diamino-2-ethoxyacridine (1:1), 7-Ethoxy-3,9-acridinediamine 2-hydroxypropanoate (1:1), 6,9-Diamino-2-ethoxyacrid
InChI Formula InChI=1S/C15H15N3O.C3H6O3/c1-2-19-10-4-6-13-12(8-10)15(17)11-5-3-9(16)7-14(11)18-13;1-2(4)3(5)6/h3-8H,2,16H2,1H3,(H2,17,18);2,4H,1H3,(H,5,6)
SMILES CCOc1ccc2nc3cc(N)ccc3c(N)c2c1.CC(O)C(=O)O
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid
Molecular Weight (g/mol) 343.377
Formula Weight 343.153