Learn More
2-ethyl-2-methyloctanoic acid, TRC
Product Code. 30337492
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337492 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30337492 Supplier Toronto Research Chemicals Supplier No. TRCB441180100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-ethyl-2-methyloctanoic acid
CAS 31199-56-1
Molecular Formula C11H22O2
Quantity 100 mg
InChI Formula InChI=1S/C11H22O2/c1-4-6-7-8-9-11(3,5-2)10(12)13/h4-9H2,1-3H3,(H,12,13)
SMILES CCCCCCC(C)(CC)C(=O)O
IUPAC Name 2-ethyl-2-methyloctanoic acid
Molecular Weight (g/mol) 186.29
Formula Weight 186.162