missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number 15446307
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
660432-43-9
C7H6FNO3
171.13
MFCD11110168
XYGZVFTWIGGORD-UHFFFAOYSA-N
2-fluoro-4-nitrophenyl methanol, 2-fluoro-4-nitrobenzyl alcohol, benzenemethanol, 2-fluoro-4-nitro, 2-fluoro-4-nitrobenzylalcohol, 2-fluoro-4-nitro-phenyl-methanol, 3-fluoro-4-hydroxymethyl nitrobenzene
17981733
(2-fluoro-4-nitrophenyl)methanol
OCC1=C(F)C=C(C=C1)[N+]([O-])=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
660432-43-9
C7H6FNO3
171.13
MFCD11110168
XYGZVFTWIGGORD-UHFFFAOYSA-N
2-fluoro-4-nitrophenyl methanol, 2-fluoro-4-nitrobenzyl alcohol, benzenemethanol, 2-fluoro-4-nitro, 2-fluoro-4-nitrobenzylalcohol, 2-fluoro-4-nitro-phenyl-methanol, 3-fluoro-4-hydroxymethyl nitrobenzene
17981733
(2-fluoro-4-nitrophenyl)methanol
OCC1=C(F)C=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Fluoro-4-nitrobenzyl alcohol
Punto di fusione 103°C to 105°C
Colore White to Yellow
Quantità 250 mg
Peso formulazione 171.1
Percent Purity 97%
Forma fisica Solid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'