missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number 15445857
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
166948-50-1
C8H3ClF4O2
242.554
MFCD16652499
VDZYQIMWGWFOEH-UHFFFAOYSA-N
2-fluoro-4-trifluoromethoxy benzoyl chloride
66523483
2-fluoro-4-(trifluoromethoxy)benzoyl chloride
C1=CC(=C(C=C1OC(F)(F)F)F)C(=O)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
166948-50-1
C8H3ClF4O2
242.554
MFCD16652499
VDZYQIMWGWFOEH-UHFFFAOYSA-N
2-fluoro-4-trifluoromethoxy benzoyl chloride
66523483
2-fluoro-4-(trifluoromethoxy)benzoyl chloride
C1=CC(=C(C=C1OC(F)(F)F)F)C(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Fluoro-4-(trifluoromethoxy)benzoyl chloride
Quantità 1 g
Numero UN UN3265
Sensibilità Moisture sensitive
Peso formulazione 242.55
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'