missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7046192
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
452-84-6
C7H8FN
125.15
MFCD00007654
QZUXMXZNVAJNSE-UHFFFAOYSA-N
3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
262970
2-fluoro-5-methylaniline
CC1=CC=C(F)C(N)=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
452-84-6
C7H8FN
125.15
MFCD00007654
QZUXMXZNVAJNSE-UHFFFAOYSA-N
3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
262970
2-fluoro-5-methylaniline
CC1=CC=C(F)C(N)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Fluoro-5-methylaniline
Densità 1.109
Punti di ebollizione 80°C to 86°C (11 mmHg)
Punto d'infiammabilità 81°C (177°F)
Odore Characteristic
Indice di rifrazione 1.533
Quantità 5 g
Numero UN UN2810
Beilstein 2715995
Peso formulazione 125.15
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'