missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzylmagnesium chloride, 0.50M in 2-MeTHF, Thermo Scientific Chemicals

Catalog Number 15421716
Click to view available options
Quantità:
100 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Fluorobenzylmagnesium chloride
CAS 120608-58-4
Formula molecolare C7H6ClFMg
Numero MDL MFCD01319900
Quantità 100 mL
Numero UN UN2924
Sensibilità Air and moisture sensitive
Sinonimo 2-fluorobenzylmagnesium chloride, chloro 2-fluorophenyl methyl magnesium, magnesium, chloro 2-fluorophenyl methyl, 2-fluorobenzylmagnesium chloride 0.25m in diethyl ether, acmc-20mp0w, 2-fluoro-benzylmagnesium chloride, kzafehnnbnglix-uhfffaoysa-m, 2-fluorobenzyl magnesium chloride, 2-fluorobenzylmagnesium chloride 0.25 m in diethyl ether, 2-fluorobenzylmagnesium chloride 0.25m solution in diethyl ether
InChI Key KZAFEHNNBNGLIX-UHFFFAOYSA-M
SMILES FC1=CC=CC=C1C[Mg]Cl
IUPAC Name magnesium;1-fluoro-2-methanidylbenzene;chloride
Molecular Weight (g/mol) 168.88
PubChem CID 2778184
Peso formulazione 168.88
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'