missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-nitropyridine, 98%, Thermo Scientific™

Catalog Number 11403368
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
6332-56-5
C5H4N2O3
140.10
MFCD00006270
BOAFCICMVMFLIT-UHFFFAOYSA-N
2-hydroxy-3-nitropyridine, 3-nitropyridin-2-ol, 3-nitro-2-pyridinol, 3-nitropyridin-2 1h-one, 2 1h-pyridinone, 3-nitro, 3-nitro-2-hydroxypyridine, pyridine, 2-hydroxy-3-nitro, 3-nitro-1,2-dihydropyridin-2-one, 3-nitro-2-oxopyridine, 2-pyridinol, 3-nitro
22793
3-nitro-1H-pyridin-2-one
[O-][N+](=O)C1=CC=CNC1=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
6332-56-5
C5H4N2O3
140.10
MFCD00006270
BOAFCICMVMFLIT-UHFFFAOYSA-N
2-hydroxy-3-nitropyridine, 3-nitropyridin-2-ol, 3-nitro-2-pyridinol, 3-nitropyridin-2 1h-one, 2 1h-pyridinone, 3-nitro, 3-nitro-2-hydroxypyridine, pyridine, 2-hydroxy-3-nitro, 3-nitro-1,2-dihydropyridin-2-one, 3-nitro-2-oxopyridine, 2-pyridinol, 3-nitro
22793
3-nitro-1H-pyridin-2-one
[O-][N+](=O)C1=CC=CNC1=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Hydroxy-3-nitropyridine
Punto di fusione ∼220°C (decomposition)
Quantità 5 g
Beilstein 124474
Peso formulazione 140.1
Percent Purity 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'