Learn More
2-Hydroxy-4-benzyloxyacetophenone, TRC
Product Code. 30407989
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30407989 Supplier Toronto Research Chemicals Supplier No. TRCH8298605MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-4-benzyloxyacetophenone
CAS 29682-12-0
Molecular Formula C15H14O3
Quantity 5 mg
Synonym Ethanone, 1-[2-hydroxy-4-(phenylmethoxy)phenyl]-, Acetophenone, 4'-(benzyloxy)-2'-hydroxy- (6CI,8CI), 1-[2-Hydroxy-4-(phenylmethoxy)phenyl]ethanone, 1-(4-(Benzyloxy)-2-hydroxyphenyl)ethan-1-one, 1-(4-Benzyloxy-2-hydroxyphenyl)ethanone, 1-[4-(Benzyloxy)-2-hydroxyphenyl]ethan-1-one, 2-Hydroxy-4-(benzyloxy)acetophenone, 2-Hydroxy-4-(phenylmethoxy)acetophenone, 2'-Hydroxy-4'-(benzyloxy)acetophenone, 4-Benzyloxy-2-hydroxy-1-acetophenone, 4-O-Benzylresacetophenone, 4'-(Benzyloxy)-2'-hydroxyacetophenone, 4'-(Benzyloxy)resacetophenone, NSC 211460, Resacetophenone 4-benzyl ether
InChI Formula InChI=1S/C15H14O3/c1-11(16)14-8-7-13(9-15(14)17)18-10-12-5-3-2-4-6-12/h2-9,17H,10H2,1H3
SMILES CC(=O)c1ccc(OCc2ccccc2)cc1O
IUPAC Name 1-(2-hydroxy-4-phenylmethoxyphenyl)ethanone
Molecular Weight (g/mol) 242.27
Formula Weight 242.0943