Learn More
2-Mercaptothiazoline, 98+%
Codice prodotto. p-7044159
Cambia vista
Click to view available options
Quantità:
100 g
500 g
Dimensione della confezione:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11321767 100 g 100g
11331767 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11321767 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L04924.22
Richiesta di formato massivo o personalizzato
Please to purchase this item. Need a web account? Register with us today!
CAS: 96-53-7 | C3H5NS2 | 119.2 g/mol
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.
Solubility
Soluble in chloroform, methanol. Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 96-53-7
Formula molecolare C3H5NS2
Molecular Weight (g/mol) 119.2
Numero MDL MFCD00126013
InChI Key WGJCBBASTRWVJL-UHFFFAOYSA-N
Sinonimo 2-mercaptothiazoline, 2-thiazolidinethione, 2-thiazoline-2-thiol, thiazolidine-2-thione, metabasal, thyroidan, 2-mercapto-2-thiazoline, mercaptothiazoline, 2-thiothiazolidone, 2-thiazolidenethione
PubChem CID 2723699
IUPAC Name 1,3-thiazolidine-2-thione
SMILES C1CSC(=S)N1
Specifica
Materiale o nome chimico 2-Mercaptothiazoline
Punto di fusione 103°C to 107°C
Quantità 100 g
Numero UN UN2811
Beilstein 106332
Informazioni di solubilità Soluble in chloroform,methanol. Slightly soluble in water.
Peso formulazione 119.21
Percent Purity ≥98%