Learn More
2-Methoxy Phenothiazine, TRC
Product Code. 30483302
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483302 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30483302 Supplier Toronto Research Chemicals Supplier No. TRCM2653301G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methoxyphenothiazine
CAS 1771-18-2
Molecular Formula C13 H11 N O S
Quantity 1 g
Synonym 2-Methoxyphenothiazine, Levomepromazine Maleate Imp. A (EP)
InChI Formula InChI=1S/C13H11NOS/c1-15-9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)16-13/h2-8,14H,1H3
SMILES COc1ccc2Sc3ccccc3Nc2c1
IUPAC Name 2-methoxy-10H-phenothiazine
Molecular Weight (g/mol) 229.3
Formula Weight 229.0561