missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-3758318
Click to view available options
Quantità:
25 g
100 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
93-25-4
C9H10O3
166.18
MFCD00004321
IVEWTCACRDEAOB-UHFFFAOYSA-N
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
7134
2-(2-methoxyphenyl)acetic acid
COC1=CC=CC=C1CC(O)=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
93-25-4
C9H10O3
166.18
MFCD00004321
IVEWTCACRDEAOB-UHFFFAOYSA-N
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
7134
2-(2-methoxyphenyl)acetic acid
COC1=CC=CC=C1CC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Methoxyphenylacetic acid
CAS min. % 98.5
CAS max % 100.0
Punto di fusione 122.0°C to 125.0°C
Colore Cream-Pink to White
Spettro a infrarossi Authentic
Intervallo percentuale saggio 99%
Imballaggio Glass bottle
Formula lineare CH3OC6H4CH2CO2H
Quantità 25 g
Beilstein 10, 188
Informazioni di solubilità Solubility in water: soluble. Other solubilities: soluble in ethenol,ethyl ether,acetone
Peso formulazione 166.18
Percent Purity 99%
Forma fisica Crystalline Powder
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'