missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Catalog Number 15444147
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
771572-43-1
C9H10F3N
189.181
MFCD06213551
WXYVXJSOJCMWGN-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethyl benzylamine, 1-2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethylbenzylamine, 2-methyl-3-trifluoromethyl phenyl methylamine
46737516
[2-methyl-3-(trifluoromethyl)phenyl]methanamine
CC1=C(C=CC=C1C(F)(F)F)CN
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
771572-43-1
C9H10F3N
189.181
MFCD06213551
WXYVXJSOJCMWGN-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethyl benzylamine, 1-2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethylbenzylamine, 2-methyl-3-trifluoromethyl phenyl methylamine
46737516
[2-methyl-3-(trifluoromethyl)phenyl]methanamine
CC1=C(C=CC=C1C(F)(F)F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Methyl-3-(trifluoromethyl)benzylamine
Indice di rifrazione 1.4812
Quantità 250 mg
Numero UN UN2735
Sensibilità Air Sensitive
Peso formulazione 189.18
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'