Learn More
2-Methyl-5-nitroimidazole-1-acetic Acid, TRC
Product Code. 30470730
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470730 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30470730 Supplier Toronto Research Chemicals Supplier No. TRCM330420100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic Acid
CAS 1010-93-1
Molecular Formula C6 H7 N3 O4
Quantity 100 mg
Synonym THIQ-3-ol isomer, 1-(3-Hydroxyphenyl)-2-methyl-1,2,3,4-tetrahydroisoquinoline-4,6-diol (1S,4R)
InChI Formula InChI=1S/C6H7N3O4/c1-4-7-2-5(9(12)13)8(4)3-6(10)11/h2H,3H2,1H3,(H,10,11)
SMILES Cc1ncc([N+](=O)[O-])n1CC(=O)O
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)acetic acid
Molecular Weight (g/mol) 185.14
Formula Weight 185.0437