missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number 15447017
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
195622-41-4
C9H7F3O3
220.147
MFCD18399742
IBHIJPBIQCIAES-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzoic acid, benzoic acid, 2-methyl-5-trifluoromethoxy
58839058
2-methyl-5-(trifluoromethoxy)benzoic acid
CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
195622-41-4
C9H7F3O3
220.147
MFCD18399742
IBHIJPBIQCIAES-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzoic acid, benzoic acid, 2-methyl-5-trifluoromethoxy
58839058
2-methyl-5-(trifluoromethoxy)benzoic acid
CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Methyl-5-(trifluoromethoxy)benzoic acid
Punto di fusione 87°C to 89°C
Colore White
Quantità 1 g
Peso formulazione 220.15
Percent Purity 97%
Forma fisica Crystalline Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'