Learn More
2-Nitro-4,5-methylenedioxybenzaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Nitro-4,5-methylenedioxybenzaldehyde |
| CAS | 712-97-0 |
| Molecular Formula | C8 H5 N O5 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C8H5NO5/c10-3-5-1-7-8(14-4-13-7)2-6(5)9(11)12/h1-3H,4H2 |
| SMILES | [O-][N+](=O)c1cc2OCOc2cc1C=O |
| IUPAC Name | 6-nitro-1,3-benzodioxole-5-carbaldehyde |
| Molecular Weight (g/mol) | 195.13 |
| Formula Weight | 195.0168 |