missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantità:
1 g
5 g
Description
Used as intermediates. Also as laboratory reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as intermediates. Also as laboratory reagents.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Used as intermediates. Also as laboratory reagents.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Materiale o nome chimico | 2-Nitro-4-(trifluoromethyl)benzyl bromide |
Punto di fusione | 36°C to 39°C |
Quantità | 1 g |
Numero UN | UN3261 |
Informazioni di solubilità | Insoluble in water. |
Peso formulazione | 284.03 |
Percent Purity | 97% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'