missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitrobenzyl alcohol, 97%

Marca:  Alfa Aesar™ A15496.36

Dettagli aggiuntivi : CAS : 612-25-9 Peso : 0.50000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantità 500g

 vedi altre versioni di questo prodotto

Codice prodotto. 11402887

  • 504.00€ / 500g


Spedizione stimata
22-04-2021
Aggiungi al carrello

Descrizione

Descrizione

Applications
The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.

Solubility
Soluble in water 5000 mg/L 20 deg Celsius.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifica

Identificatori chimici

612-25-9
153.137
BWRBVBFLFQKBPT-UHFFFAOYSA-N
11923
C1=CC=C(C(=C1)CO)[N+](=O)[O-]
C7H7NO3
MFCD00007186
2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
(2-nitrophenyl)methanol

Specifica

2-Nitrobenzyl alcohol
270°C
500g
C7H7NO3
2046649
Soluble in water 5000mg/L 20 deg Celsius.
C1=CC=C(C(=C1)CO)[N+](=O)[O-]
153.137
153.14
Light sensitive
70°C to 74°C
612-25-9
MFCD00007186
2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
BWRBVBFLFQKBPT-UHFFFAOYSA-N
(2-nitrophenyl)methanol
11923
97%
Videos
SDS Documenti