Learn More
2-Phenylquinazolin-4(3h)-one, TRC
Product Code. 30421246
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421246 100 mg -
30411979 10 mg 10mg
30420854 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30421246 Supplier Toronto Research Chemicals Supplier No. TRCP399718100MG
 Restricted Item
Spedizione stimata: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1022-45-3
Molecular Formula C14H10N2O
Molecular Weight (g/mol) 222.24
Synonym 2-Phenyl-4(3H)-quinazolinone
IUPAC Name 2-phenylquinazolin-4-ol
SMILES Oc1nc(nc2ccccc12)c3ccccc3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H10N2O/c17-14-11-8-4-5-9-12(11)15-13(16-14)10-6-2-1-3-7-10/h1-9H,(H,15,16,17)
Formula Weight 222.0793
Chemical Name or Material 2-Phenylquinazolin-4(3h)-one