Learn More
Tocris Bioscience™ 2-Phospho-L-ascorbic acid trisodium salt
Ascorbic acid derivative; maintains differentiation potential in bone marrow-derived MSCs
Marca: Tocris Bioscience™ 5778/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
2-Phospho-L-ascorbic acid trisodium salt is a stable ascorbic acid derivative used in cell culture; in combination with FGF-2, maintains differentiation potential in bone marrow-derived mesenchymal stem cells (MSC) through increased expression of HGF. Also exhibits synergistic protection of hMSCs under oxidative stress in combination with N-acetylcysteine (Cat. No. 5619).Specifica
| 2-(Dihydrogen phosphate)-L-ascorbic acid sodium salt | |
| 66170-10-3 | |
| 50 mg |