missing translation for 'onlineSavingsMsg'
Learn More

2-Propyl-1-heptanol, TRC

Codice prodotto. 30137259
Cambia vista
Click to view available options
Quantità:
1 g
5 g
Dimensione della confezione:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30137259 5 g 5g
17982495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137259 Supplier Toronto Research Chemicals Supplier No. TRCP8358205G
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-59-8
Formula molecolare C10 H22 O
Molecular Weight (g/mol) 158.28
Sinonimo 1-Heptanol, 2-propyl- (6CI, 7CI, 8CI, 9CI, ACI), 2-Propyl-1-heptanol (ACI), 2-Propylheptanol, 2-Propylheptyl alcohol
IUPAC Name 2-propylheptan-1-ol
SMILES CCCCCC(CO)CCC

Specifications

Materiale o nome chimico 2-Propyl-1-heptanol
Quantità 5 g
InChI Formula InChI=1S/C10H22O/c1-3-5-6-8-10(9-11)7-4-2/h10-11H,3-9H2,1-2H3
Peso formulazione 158.1671
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.