Learn More
2-Quinoxalinecarboxaldehyde 1,4-Dioxide Dimethyl Acetal, TRC
Specifica
Specifica
| Materiale o nome chimico | 2-Quinoxalinecarboxaldehyde 1,4-Dioxide Dimethyl Acetal |
| CAS | 32065-66-0 |
| Formula molecolare | C14 H9 Cl O3 |
| Quantità | 50 mg |
| Sinonimo | 2-Formylquinoxaline 1,4-Dioxide Dimethyl Acetal, Quinoxaline-2-carboxaldehyde Dimethyl aAetal Di-N-oxide |
| InChI Formula | InChI=1S/C14H9ClO3/c15-13-6-5-9(7-12(13)14(17)18)11-4-2-1-3-10(11)8-16/h1-8H,(H,17,18) |
| SMILES | OC(=O)c1cc(ccc1Cl)c2ccccc2C=O |
| IUPAC Name | 2-chloro-5-(2-formylphenyl)benzoic acid |
| Molecular Weight (g/mol) | 260.673 |
| Peso formulazione | 260.024 |