Learn More
Dinoseb, SPEX CertiPrep™
Codice prodotto. 12959204 Sfoglia Tutto SPEX Certiprep Prodotti
Cambia vista
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
12959204 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 12959204 Fornitore SPEX Certiprep N. del fornitore S1710
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Dinoseb88-85-71000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifica
Materiale o nome chimico Dinoseb
Imballaggio 2mL amber prescored glass ampule
CAS 88-85-7, 67-56-1
CAS min. % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS max % 0.1, 99.9
Formula molecolare C10H12N2O5
Da utilizzare con (apparecchiatura) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key OWZPCEFYPSAJFR-UHFFFAOYSA-N
SMILES CCC(C)C1=CC(=CC(=C1O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2-butan-2-yl-4,6-dinitrophenol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 240.215
PubChem CID 6950
Quantità 1 mL
ChEBI CHEBI:83632
Product intended for laboratory use only