missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanone, 98%, Thermo Scientific Chemicals

Catalog Number p-7023773
Click to view available options
Quantità:
100 g
500 g
2500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
112-12-9
C11H22O
170.30
MFCD00009583
KYWIYKKSMDLRDC-UHFFFAOYSA-N
2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
8163
CHEBI:17700
undecan-2-one
CCCCCCCCCC(C)=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
112-12-9
C11H22O
170.30
MFCD00009583
KYWIYKKSMDLRDC-UHFFFAOYSA-N
2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
8163
CHEBI:17700
undecan-2-one
CCCCCCCCCC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Undecanone
Punto di fusione 11°C to 13°C
Punti di ebollizione 228°C
Punto d'infiammabilità 89°C (192°F)
Quantità 100 g
Numero UN UN3082
Beilstein 1749573
Indice di Merck 14,6104
Peso formulazione 170.3
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'