Learn More
20-Deoxo-20-(1-piperidinyl)tylosin, TRC
Product Code. 30351880
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351880 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30351880 Supplier Toronto Research Chemicals Supplier No. TRCD23151010MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 20-Deoxo-20-(1-piperidinyl)tylosin
CAS 1003024-00-7
Molecular Formula C51H88N2O16
Quantity 10 mg
InChI Formula InChI=1S/C51H88N2O16/c1-13-38-35(27-63-50-47(62-12)46(61-11)42(57)31(5)65-50)23-28(2)17-18-36(54)29(3)24-34(19-22-53-20-15-14-16-21-53)44(30(4)37(55)25-39(56)67-38)69-49-43(58)41(52(9)10)45(32(6)66-49)68-40-26-51(8,60)48(59)33(7)64-40/h17-18,23,29-35,37-38,40-50,55,57-60H,13-16,19-22,24-27H2,1-12H3/b18-17+,28-23+/t29-,30+,31-,32-,33+,34+,35-,37-,38-,40+,41-,42-,43-,44-,45-,46?,47?,48+,49+,50-,51-/m1/s1
SMILES CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CCN4CCCCC4)C[C@@H](C)C(=O)\C=C\C(=C\[C@@H]1CO[C@@H]5O[C@H](C)[C@@H](O)C(OC)C5OC)\C
IUPAC Name (4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxymethyl]-5,9,13-trimethyl-7-(2-piperidin-1-ylethyl)-1-oxacyclohexadeca-11,13-diene-2,10-dione
Molecular Weight (g/mol) 985.25
Formula Weight 984.6134