Learn More
Greiner Bio-One 20 μL Universal Sterile Filter Tips
Prevents contamination with liquids and aerosols during pipetting process.
Marca: Greiner Bio-One 774288
Dettagli aggiuntivi : Peso : 1.33000kg
Descrizione
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Non-cytotoxic
- Prevents aspiration of particles into interior of pipette
- Minimizes danger of unwanted cross-contamination
- Works with DNA and RNA and ra
Specifica
20 μL | |
Sterile | |
Irradiation | |
Costar™ Onepette™, Eppendorf™ Reference™, Gilson™ Pipetman™ P20, Socorex™ Acura™ Calibra | |
Polypropylene |
Filtered | |
Translucent | |
Universal | |
No |