Learn More
Thermo Scientific™ 24-Position Sample Tube Rack
Product Code. 17865372 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Product Code. 17865372 Supplier Thermo Scientific™ Supplier No. 942347003911
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Customize your Thermo Scientific™ CETAC™ Autosampler with a range of CETAC™ Autosampler Sample Tube Racks to suit your requirements.
Match Your CETAC Sample Tube Rack to Your Laboratory’s Needs
  • 21-position sample tube rack
  • 24-position sample tube rack
  • 40-position sample tube rack
  • 60-position sample tube rack
  • 90-position sample tube rack
  • Extended Rack for CETAC ASX-520 Autosampler (for use with Thermo Scientific™ iCAP™ Q Series ICP-MS systems)
Specifications
For Use With (Equipment) CETAC, ESI
Type Autosampler Sample Tube Racks
Includes 24-Position Sample Tube Rack
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.