Learn More
(2S)-2-Amino-hexanedioic Acid, TRC
Product Code. 17993175
Change view
Click to view available options
Quantity:
250 mg
500 mg
5 g
Unit Size:
250mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993175 250 mg 250mg
17931905 500 mg 500mg
17901475 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993175 Supplier Toronto Research Chemicals Supplier No. TRCA609970250MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1118-90-7
Molecular Formula C6 H11 N O4
Synonym (2S)-2-Aminohexanedioic Acid∣Hexanedioic acid, 2-amino-, (S)-∣Hexanedioic acid, 2-amino-, L- (8CI)∣(2S)-2-Aminohexanedioic acid∣(S)-2-Aminoadipic acid∣(S)-2-Aminohexanedioic acid∣L-2-Aminoadipic acid∣L-Aminoadipic acid∣L-α-Aminoadipate∣L-α-Aminoadipic acid
Specifications
Quantity 250 mg
Shipping Condition Room Temperature
Formula Weight 161.0688
Molecular Weight (g/mol) 161.16
Chemical Name or Material L-2-Aminoadipic acid