Learn More
(2S,4E,6S)-2,4,6-Trimethyl-7-oxo-7-[(4S)-2-oxo-4-(phenylmethyl)-3-oxazolidinyl]-4-heptenoic Acid Ethyl Ester, TRC
Codice prodotto. 30450035
Cambia vista
Click to view available options
Quantità:
500 mg
Dimensione della confezione:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30450035 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30450035 Fornitore Toronto Research Chemicals N. del fornitore TRCT796680500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico (2S,4E,6S)-2,4,6-Trimethyl-7-oxo-7-[(4S)-2-oxo-4-(phenylmethyl)-3-oxazolidinyl]-4-heptenoic Acid Ethyl Ester
CAS 943858-30-8
Formula molecolare C22H29NO5
Quantità 500 mg
InChI Formula InChI=1S/C22H29NO5/c1-5-27-21(25)17(4)12-15(2)11-16(3)20(24)23-19(14-28-22(23)26)13-18-9-7-6-8-10-18/h6-11,16-17,19H,5,12-14H2,1-4H3/b15-11+/t16-,17-,19+/m0/s1
SMILES CCOC(=O)[C@@H](C)C\C(=C\[C@H](C)C(=O)N1[C@H](Cc2ccccc2)COC1=O)\C
IUPAC Name ethyl (E,2S,6S)-7-[(4R)-4-benzyl-2-oxo-1,3-oxazolidin-3-yl]-2,4,6-trimethyl-7-oxohept-4-enoate
Molecular Weight (g/mol) 387.47
Peso formulazione 387.2046