Learn More
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane, 99%
Product Code. 15441337
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15441337 Supplier Thermo Scientific Alfa Aesar Supplier No. 033668.03
 Restricted Item
In stock
Specifications
Chemical Name or Material (2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane
CAS 77876-39-2
Molecular Formula C29H30P2
MDL Number MFCD00058923
Quantity 1 g
Synonym 2s,4s---2,4-bis diphenylphosphino pentane, s,s-bdpp, 2s,4s-pentane-2,4-diylbis diphenylphosphine, phosphine, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenyl, s,s-2,4-bis diphenylphosphino pentane, 2s,4s-2,4-bis diphenylphosphino pentane, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenylphosphine, 2s,4s-4-diphenylphosphanylpentan-2-yl-diphenylphosphane, 2s,4s-4-diphenylphosphanyl pentan-2-yl diphenylphosphane
InChI Key CTYPJIUQROQJBG-DQEYMECFSA-N
SMILES CC(CC(C)P(C1=CC=CC=C1)C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4
IUPAC Name [(2S,4S)-4-diphenylphosphanylpentan-2-yl]-diphenylphosphane
Molecular Weight (g/mol) 440.507
PubChem CID 2734567
Formula Weight 440.5
Percent Purity 99%
Physical Form Solid